Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163051.1 | NP_001156523.1 | 4 - 5 | 1152 | 88 | |
GenBank mRNA | FJ958366.1 | - | 4 - 5 | 1152 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |