Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009739.1 | NP_001009739.1 | 6 - 7 | 1197 | 109 | |
GenBank mRNA | AF227759.1 | - | 6 - 7 | 1197 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |