Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009792.2 | NP_001009792.1 | 4 - 5 | 451 | 130 | |
XM_012101545.3 | XP_011956935.1 | 4 - 5 | 488 | 130 | ||
GenBank mRNA | U94789.1 | - | 4 - 5 | 451 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |