Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205151.1 | NP_001192080.1 | 1 | 120 | 113 | |
GenBank mRNA | HQ263381.1 | - | 1 | 120 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |