Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959276.1 | XP_027815077.1 | 8 - 9 | 1806 | 60 | |
XM_027959277.1 | XP_027815078.1 | 8 - 9 | 3878 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |