Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102002.2 | XP_014957488.2 | 2 - 3 | 3924 | 116 | |
XM_027957236.1 | XP_027813037.1 | 2 - 3 | 4082 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |