Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971079.1 | XP_027826880.1 | 2 - 3 | 475 | 104 | |
XM_027971080.1 | XP_027826881.1 | 2 - 3 | 475 | 104 | ||
XM_027971082.1 | XP_027826883.1 | 2 - 3 | 204 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |