Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959954.1 | XP_027815755.1 | 10 - 11 | 1534 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |