Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012180072.3 | XP_012035462.1 | 5 - 6 | 783 | 85 | |
XM_012180074.3 | XP_012035464.1 | 5 - 6 | 741 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |