Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101107932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963378.1 | XP_027819179.1 | 3 - 4 | 1529 | 66 |
Gene Symbol: LOC114111613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963438.1 | XP_027819239.1 | 3 - 4 | 1529 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |