Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965894.1 | XP_027821695.1 | 4 - 5 | 456 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |