Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014925.3 | XP_004014974.1 | 2 - 3 | 282 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |