Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIF4GD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974065.1 | XP_027829866.1 | 2 - 3 | 223 | 94 | |
XM_027974066.1 | XP_027829867.1 | 2 - 3 | 209 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |