Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105611393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962507.1 | XP_027818308.1 | 2 - 3 | 251 | 88 |
Gene Symbol: LOC114110955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962512.1 | XP_027818313.1 | 2 - 3 | 538 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |