Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972699.1 | XP_027828500.1 | 5 - 6 | 904 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |