Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962301.1 | XP_027818102.1 | 3 - 4 | 508 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |