Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006348.4 | XP_004006397.2 | 3 - 4 | 692 | 73 | |
XM_027967315.1 | XP_027823116.1 | 3 - 4 | 1303 | 73 | ||
XM_027967316.1 | XP_027823117.1 | 4 - 5 | 865 | 73 | ||
XM_027967317.1 | XP_027823118.1 | 2 - 3 | 1237 | 73 | ||
XM_027967318.1 | XP_027823119.1 | 2 - 3 | 787 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |