Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012180966.3 | XP_012036356.1 | 1 - 2 | 213 | 88 | |
XM_012180967.3 | XP_012036357.1 | 1 - 2 | 202 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |