Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016413.3 | XP_004016462.1 | 4 - 5 | 498 | 162 | |
XM_012096242.3 | XP_011951632.1 | 4 - 5 | 522 | 162 | ||
XM_012096243.3 | XP_011951633.1 | 6 - 7 | 632 | 162 | ||
XM_027979635.1 | XP_027835436.1 | 5 - 6 | 574 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |