Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016999.3 | XP_004017048.2 | 2 - 3 | 226 | 127 | |
XM_012096943.2 | XP_011952333.2 | 2 - 3 | 228 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |