Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975703.1 | XP_027831504.1 | 9 - 10 | 2058 | 89 | |
XM_027975704.1 | XP_027831505.1 | 11 - 12 | 1995 | 89 | ||
XM_027975705.1 | XP_027831506.1 | 10 - 11 | 1968 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |