Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020570.4 | XP_004020619.1 | 2 - 3 | 376 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |