Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103654.2 | XP_014959140.2 | 2 - 3 | 331 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |