Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012181035.3 | XP_012036425.1 | 3 - 4 | 2315 | 107 | |
XM_015096904.2 | XP_014952390.1 | 1 - 2 | 1165 | 107 | ||
XM_015096905.2 | XP_014952391.1 | 3 - 4 | 2167 | 107 | ||
XM_027970077.1 | XP_027825878.1 | 2 - 3 | 1020 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |