Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963488.1 | XP_027819289.1 | 1 - 2 | 460 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |