Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195723.1 | NP_001182652.1 | 1 - 2 | 371 | 59 | |
GenBank mRNA | AF042497.1 | - | 1 - 2 | 371 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Inventoried Probe spans exons |