Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017343835.1 | XP_017199324.1 | 3 - 4 | 1191 | 61 | |
GenBank mRNA | AY603960.1 | - | 2 - 3 | 927 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |