Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082005.1 | NP_001075474.1 | 4 - 5 | 568 | 71 | |
XM_008254828.2 | XP_008253050.1 | 4 - 5 | 1166 | 71 | ||
XM_008254829.2 | XP_008253051.1 | 4 - 5 | 1167 | 71 | ||
XM_017340967.1 | XP_017196456.1 | 4 - 5 | 1167 | 71 | ||
XM_017340968.1 | XP_017196457.1 | 4 - 5 | 1168 | 71 | ||
XM_017340969.1 | XP_017196458.1 | 4 - 5 | 1167 | 71 | ||
XM_017340970.1 | XP_017196459.1 | 4 - 5 | 1168 | 71 | ||
XM_017340971.1 | XP_017196460.1 | 3 - 4 | 1053 | 71 | ||
XM_017340972.1 | XP_017196461.1 | 4 - 5 | 1168 | 71 | ||
XM_017340973.1 | XP_017196462.1 | 4 - 5 | 1170 | 71 | ||
XM_017340974.1 | XP_017196463.1 | 3 - 4 | 323 | 71 | ||
XM_017340975.1 | XP_017196464.1 | 4 - 5 | 1170 | 71 | ||
GenBank mRNA | AB038263.1 | - | 4 - 5 | 568 | 71 | |
AB038264.1 | - | 4 - 5 | 568 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |