Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082207.1 | NP_001075676.1 | 2 - 3 | 421 | 104 | |
XM_017342615.1 | XP_017198104.1 | 2 - 3 | 455 | 104 | ||
GenBank mRNA | D49841.1 | - | 2 - 3 | 421 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |