Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082713.1 | NP_001076182.1 | 554 | 100 | ||
GenBank mRNA | AF091738.1 | - | 554 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus |