Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082787.2 | NP_001076256.1 | 3 - 4 | 400 | 103 | |
XM_008255647.2 | XP_008253869.1 | 4 - 5 | 595 | 103 | ||
GenBank mRNA | AY101395.1 | - | 3 - 4 | 402 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |