Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC28A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082791.1 | NP_001076260.1 | 11 - 12 | 1116 | 102 | |
GenBank mRNA | AF161716.1 | - | 11 - 12 | 1116 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |