Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082632.1 | NP_001076101.1 | 1 | 247 | 102 | |
GenBank mRNA | X78139.1 | - | 1 | 122 | 102 | |
Z67878.1 | - | 1 | 247 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Inventoried Both primers and probe map within a single exon |