Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082790.1 | NP_001076259.1 | 1 | 851 | 59 | |
GenBank mRNA | X89731.1 | - | 851 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |