Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163067.1 | NP_001156539.1 | 1 - 2 | 313 | 88 | |
XM_017350167.1 | XP_017205656.1 | 2 - 3 | 303 | 88 | ||
GenBank mRNA | AY165034.1 | - | 1 - 2 | 313 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |