Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTEX1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092984.4 | XP_001092984.2 | 1655 | 116 | ||
GenBank mRNA | JU482003.1 | - | 259 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |