Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194746.3 | NP_001181675.1 | 447 | 71 | ||
GenBank mRNA | JU320622.1 | - | 447 | 71 | ||
JU471566.1 | - | 393 | 71 | |||
JV048624.1 | - | 283 | 71 | |||
JV486254.1 | - | 229 | 71 | |||
JV632743.1 | - | 228 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |