Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015132681.2 | XP_014988167.1 | 1417 | 64 | ||
XM_015132682.2 | XP_014988168.1 | 763 | 64 | |||
XM_028845211.1 | XP_028701044.1 | 1414 | 64 | |||
GenBank mRNA | JV630757.1 | - | 445 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |