Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM219B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194738.2 | NP_001181667.2 | 375 | 82 | ||
XM_015142708.2 | XP_014998194.2 | 805 | 82 | |||
XM_015142710.2 | XP_014998196.2 | 805 | 82 | |||
XM_015142711.2 | XP_014998197.2 | 805 | 82 | |||
GenBank mRNA | JU322524.1 | - | 414 | 82 | ||
JV638133.1 | - | 336 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |