Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194402.1 | NP_001181331.1 | 1687 | 55 | ||
XM_015148849.2 | XP_015004335.1 | 1237 | 55 | |||
XM_015148850.2 | XP_015004336.1 | 1332 | 55 | |||
XM_028827416.1 | XP_028683249.1 | 6695 | 55 | |||
GenBank mRNA | JU335680.1 | - | 1299 | 55 | ||
JU472238.1 | - | 1214 | 55 | |||
JV047779.1 | - | 1241 | 55 | |||
JV720045.1 | - | 1211 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |