Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261558.1 | NP_001248487.1 | 1318 | 96 | ||
XM_015144304.2 | XP_014999790.1 | 1516 | 96 | |||
XM_028850190.1 | XP_028706023.1 | 1253 | 96 | |||
XM_028850191.1 | XP_028706024.1 | 4501 | 96 | |||
GenBank mRNA | JU320570.1 | - | 1046 | 96 | ||
JU335058.1 | - | 1318 | 96 | |||
JV717167.1 | - | 1255 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |