Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266749.1 | NP_001253678.1 | 965 | 96 | ||
XM_015147324.2 | XP_015002810.1 | 1024 | 96 | |||
XM_015147325.2 | XP_015002811.1 | 1089 | 96 | |||
XM_015147326.2 | XP_015002812.1 | 979 | 96 | |||
XM_015147327.2 | XP_015002813.1 | 960 | 96 | |||
XM_015147328.2 | XP_015002814.1 | 1044 | 96 | |||
XM_028853261.1 | XP_028709094.1 | 885 | 96 | |||
XM_028853262.1 | XP_028709095.1 | 866 | 96 | |||
XM_028853263.1 | XP_028709096.1 | 950 | 96 | |||
XM_028853264.1 | XP_028709097.1 | 840 | 96 | |||
XM_028853265.1 | XP_028709098.1 | 821 | 96 | |||
GenBank mRNA | JU320257.1 | - | 984 | 96 | ||
JU334281.1 | - | 920 | 96 | |||
JU334282.1 | - | 965 | 96 | |||
JV649904.1 | - | 1161 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |