Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261137.1 | NP_001248066.1 | 497 | 61 | ||
GenBank mRNA | JU322182.1 | - | 490 | 61 | ||
JU473766.1 | - | 497 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |