Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260998.1 | NP_001247927.1 | 305 | 71 | ||
GenBank mRNA | JU333224.1 | - | 305 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta Inventoried |