Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194359.1 | NP_001181288.1 | 1090 | 57 | ||
XM_028840529.1 | XP_028696362.1 | 1126 | 57 | |||
GenBank mRNA | JU323478.1 | - | 906 | 57 | ||
JV495664.1 | - | 975 | 57 | |||
JV534235.1 | - | 925 | 57 | |||
JV600241.1 | - | 940 | 57 | |||
JV643048.1 | - | 1064 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |