Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257556.1 | NP_001244485.1 | 828 | 63 | ||
GenBank mRNA | JU320923.1 | - | 828 | 63 | ||
JU473059.1 | - | 804 | 63 | |||
JV646005.1 | - | 426 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |