Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261521.1 | NP_001248450.1 | 369 | 104 | ||
XM_015138520.2 | XP_014994006.1 | 964 | 104 | |||
XM_015138521.2 | XP_014994007.1 | 591 | 104 | |||
XM_015138522.2 | XP_014994008.1 | 398 | 104 | |||
GenBank mRNA | JU322866.1 | - | 369 | 104 | ||
JU322868.1 | - | 369 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |