Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257787.1 | NP_001244716.1 | 2289 | 69 | ||
GenBank mRNA | JU320203.1 | - | 2289 | 69 | ||
JU471441.1 | - | 2233 | 69 | |||
JV043937.1 | - | 2355 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |