Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3HDML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001081974.4 | XP_001081974.2 | 974 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |