Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260895.2 | NP_001247824.1 | 1200 | 71 | ||
XM_015131601.2 | XP_014987087.1 | 2087 | 71 | |||
XM_028843157.1 | XP_028698990.1 | 1193 | 71 | |||
GenBank mRNA | JV045626.1 | - | 1202 | 71 | ||
JV704315.1 | - | 1271 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |